5-Amino-1H-tetrazole monohydrate 99%
Leverandør: Thermo Scientific
|
Danger
|
Synonymer:
5-Aminotetrazole monohydrate, 5-Amino-1H-tetrazole monohydrate, Tetrazol-5-amine, 1H-Tetrazol-5-ylamine monohydrate, 1H-Tetrazol-5-amine hydrate, 5-Amino-tetrazole-monohydrate
B24018.14
B24018.22
B24018.36
B24018.14EA
395
NOK
B24018.14
B24018.22
B24018.36
5-Amino-1H-tetrazole monohydrate 99%
5-Amino-1H-tetrazole monohydrate
Purity: 99%
CAS No.: 15454-54-3
Molecular Formula: CH3N5.H2O
Formula Weight: 103.08 (85.06anhy)
pH: 3.7
Melting Point: ca 203° dec.
Beilstein Reference: 108872
UN No.: 1325
MDL No.: MFCD00149327
Formel:
CH₃N₅·1H₂O MW: 103,08 g/mol Smeltepunkt: ∼ 203 °C (dec.) Tetthet: 1,51 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00149327 CAS-nummer: 15454-54-3 EINECS: 224-581-7 FN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...