1-Butyl-3-methylimidazolium bromide ≥99%
Leverandør: Thermo Scientific
Warning
Synonymer: 3-Butyl-1-methyl-1H-imidazol-3-ium bromide
H27201.18 H27201.06
H27201.18EA 1850 NOK
H27201.18 H27201.06
1-Butyl-3-methylimidazolium bromide ≥99%
1-Butyl-3-methylimidazolium bromide
Purity: 99% CAS No.: 85100-77-2 Molecular Formula: C8H15BrN2 Formula Weight: 219.12 Melting Point: 65-75° MDL No.: MFCD03427611
Formel: C₈H₁₅BrN₂
MW: 219,12 g/mol
Smeltepunkt: 65…75 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03427611
CAS-nummer: 85100-77-2