Pentyl-4-hydroxybenzoate ≥98%
Leverandør: Thermo Scientific
Synonymer: Pentylparaben, 4-Hydroxybenzoic acid Amyl Ester, 4-Hydroxybenzoic acid pentyl ester, Pentyl parahydroxybenzoate, Pentyl 4-Hydroxybenzoate, 4-Hydroxybenzoic acid Pentyl Ester, Amyl 4-Hydroxybenzoate, Amylparaben, n-Pentyl 4-hydroxybenzoate
L05563.06 L05563.14
L05563.06EA 690 NOK
L05563.06 L05563.14
Pentyl-4-hydroxybenzoate ≥98%
Pentyl 4-hydroxybenzoate
Purity: 98% CAS No.: 6521-29-5 Molecular Formula: C12H16O3 Formula Weight: 208.26 Melting Point: 36-40° Boiling Point: 197-200°/10mm MDL No.: MFCD00016479
Formel: C₁₂H₁₆O₃
MW: 208,26 g/mol
Kokepunkt: 197…200 °C (10 mmHg)
Smeltepunkt: 36…40 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016479
CAS-nummer: 6521-29-5
EINECS: 229-408-9