trans-1,2-sykloheksandikarboksyl syre ≥98%
Leverandør: Thermo Scientific
Warning
Synonymer: trans-Cyclohexane-1,2-dicarboxylic acid
L11288.06 L11288.14
L11288.06EA 810 NOK
L11288.06 L11288.14
trans-1,2-sykloheksandikarboksyl syre ≥98%
trans-1,2-sykloheksandikarboksyl syre
Purity: 98% CAS No.: 2305-32-0 Molecular Formula: C8H12O4 Formula Weight: 172.18 Melting Point: 220-221° Beilstein Reference: 3200609
Formel: C₈H₁₂O₄
MW: 172,18 g/mol
Kokepunkt: 339 °C (1013 hPa)
Smeltepunkt: 220…221 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001464
CAS-nummer: 2305-32-0
EINECS: 218-975-8