Pentaerytritol, Sigma-Aldrich®

Leverandør: Merck


M-Clarity™ quality level = MQ200

8.07331.0100 8.07331.1000
8.07331.0100EA 368 NOK
8.07331.0100 8.07331.1000
Pentaerytritol, Sigma-Aldrich®
Pentaerytritol
Formel: C₅H₁₂O₄
MW: 136,15 g/mol
Kokepunkt: 380 °C (1013 hPa)
Smeltepunkt: 260…264 °C
Tetthet: 1,511 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004692
CAS-nummer: 115-77-5
EINECS: 204-104-9
Merck Indeks: 12,07245

Bestill nå

Spesifikasjon Testresultater

Solubility 72.3 g/l (25 °C)
Melting Point 256 - 258 °C
Molar Mass 136.14 g/mol
Bulk Density 750 - 850 kg/m3
Boiling Point 276 °C (40 hPa)
Vapor Pressure  - 0.00001 hPa (20 °C)
Density 1.39 g/cm3 (20 °C)
pH 3.5 - 4.5 (100 g/l, H2O, 35 °C)
Ignition Point 490 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR