Benzyltrimethylammonium bromide
Leverandør: Molekula
|
Warning
|
MOLE18152625-100GEA
0
NOK
MOLE18152625-100G
Benzyltrimethylammonium bromide
Benzyltrimethylammonium bromide
Formel:
C₁₀H₁₆BrN MW: 230,15 g/mol Smeltepunkt: 235 °C |
MDL nummer:
MFCD00011780 CAS-nummer: 5350-41-4 EINECS: 226-320-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...