(Methoxymethyl)triphenylphosphonium chloride
Leverandør: Molekula
|
Warning
|
MOLEM64295631EA
1600
NOK
MOLEM64295631
(Methoxymethyl)triphenylphosphonium chloride
(Methoxymethyl)triphenylphosphonium chloride
Formel:
C₂₀H₂₀ClOP MW: 342,8 g/mol Smeltepunkt: 190 °C |
MDL nummer:
MFCD00011800 CAS-nummer: 4009-98-7 EINECS: 223-664-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...