Dibenzothiophene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Dibenzo[b,d]thiophene

D0148-25G D0148-5G
TCIAD0148-25GEA 1100 NOK
TCIAD0148-25G TCIAD0148-5G
Dibenzothiophene ≥98.0% (by GC)
Dibenzothiophene
Formel: C₁₂H₈S
MW: 184,26 g/mol
Kokepunkt: 333°C
Smeltepunkt: 95…98 °C
Tetthet: 1,1295 g/cm³ (125 °C)
Flash punkt: 170 °C
MDL nummer: MFCD00004969
CAS-nummer: 132-65-0
EINECS: 205-072-9
FN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a05c133bd227f41cad306702621564dcdc50394&vid=9a05c133bd227f41cad306702621564dcdc50394&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 97.0 to 101.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR