3-metoksy-1-butanol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
3-Methoxybutan-1-ol
M0109-25ML
M0109-500ML
TCIAM0109-25MLEA
220
NOK
TCIAM0109-25ML
TCIAM0109-500ML
3-metoksy-1-butanol ≥98.0% (by GC)
3-metoksy-1-butanol
Formel:
C₅H₁₂O₂ MW: 104,15 g/mol Kokepunkt: 158°C Smeltepunkt: –85 °C Tetthet: 0,92 g/cm³ (20 °C) Flash punkt: 64°C |
MDL nummer:
MFCD00002931 CAS-nummer: 2517-43-3 EINECS: 219-741-8 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...