(3-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-(3-Nitrophenoxy)acetic acid, 3-Nitrophenoxyacetic acid

N0224-25G N0224-5G
TCIAN0224-25GEA 2450 NOK
TCIAN0224-25G TCIAN0224-5G
(3-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(3-Nitrophenoxy)acetic acid
Formel: (C₆H₄(NO₂))OCH₂COOH
MW: 197,15 g/mol
MDL nummer: MFCD00059826
CAS-nummer: 1878-88-2
EINECS: 217-528-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d22acabb8e20a40e3cac5977de7040d252e87b9c&vid=d22acabb8e20a40e3cac5977de7040d252e87b9c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 151.0 to 155.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR