Stirling engine, transparent
Leverandør: PHYWE
04372.00 04371.00 04371.97 04372.01 04372.02 04372.03 04372.04 32154.00
765-1183EA 38025 NOK
765-1183 765-1181 765-1182 765-1184 765-1185 765-1186 765-1187 765-1330
Stirling engine, transparent
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Phywe's transparent Stirling engine clearly shows the engine's components and operation. For the demonstration of the function of.
  • Hot air engine.
  • Heat pump / cooling machine.
  • Solar motor
For determination of: Mechanical and electrical power as a function of speed. Operating temperatures and temperature differences. Efficiency. For measurement and evaluation of pV diagrams.
Stirling engine, transparent: Stirling engine mounted on lacquered base plate. Large transparent flywheel. Cylinder and piston of heat-resistant glass. Crankshaft and shaft madeMore Product Information