Measurement inserts for resistance measurement
Leverandør: PHYWE
11109.01 11109.02
765-1291EA 11700 NOK
765-1291 765-1292
Measurement inserts for resistance measurement
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
Measurement insert for resistance measurement. Used together with demonstration instruments 765-1247. Clear, fine-grained scale with number markings. Measuring range window where the information automatically follows the measuring range switch. On the back there are measuring range switches, zero adjustment and contact sleeves for printers.