Experiment kit, electric field lines
Leverandør: PHYWE
06251.88 06251.01 06251.02 06251.03 06251.04 06251.05 06251.06
765-1199EA 15000 NOK
765-1199 765-1193 765-1194 765-1195 765-1196 765-1197 765-1198
Experiment kit, electric field lines
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
For demonstration of electric fields between electrodes of different shapes. The electrodes have been applied in thin conductive layers on plexiglass plates and the field line images appear in groats placed in a cuvette with 1 to 2 mm castor oil (the cuvette is placed on the plexiglass plate). The cuvette has an annular electrode in the outer jacket and a 4 mm connection sleeve. The cleaning is easy when electrodes resp. plexiglass plate never comes into contact with the oil. Suitable voltage source: High voltage generator 0 to 25 kV or Van de Graff generator.
  • Dimensions: plates 180 × 1