Conductor balls
Leverandør: PHYWE
06236.00 06237.00 06238.00
765-1190EA 1475 NOK
765-1190 765-1191 765-1192
Conductor balls
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
For electrostatic experiments, for example, determination of the charge of a sphere and induction experiments. Hollow surface-treated brass sphere equipped with 4 mm holes for connecting banana connectors.