Rubber belts and drive belts for generators
Leverandør: GR NNA PLASTEMBALLAGE AB
194765-145 R11-01540
765-0934EA 0 NOK
765-0934 765-0325
Rubber belts and drive belts for generators
Lærings materiell Læremateriale for fysikk Electricity/Magnetism Learning Activities
765-0325 Rubber band, Norstedt; Fits Norstedt's belt generators.
  • Dimensions: 100 × 7 cm
765-0934 Drive belt, Norstedt, 12 V; Fits Norstedt's belt generators with 12 V motor
765-1354 Drive belt, Norstedt, 220 V; Fits Norstedt's hand-operated belt generators and those with a 220 V motor