Magnetic field, force
Leverandør: PHYWE
06094.00 06410.00 06412.00 06414.00
765-1188EA 0 NOK
765-1188 765-1200 765-1201 765-1202
Magnetic field, force
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Used in studies of the power action of a current-carrying conductor in a magnetic field. The field can e.g. obtained from U-magnet 765-0444. With two bands, the force action between these at different current directions can be investigated. Each band is provided at both ends with a 4 mm banana plug.
  • Metal band, 2 pcs.
  • Dimensions: 6 × 900 mm
Konduktorgunga. For demonstration of the force exerted by a magnetic field on a conductor through which a current passes. Aluminum tube attached to two bendable woven metal strips. Each end has a 4 mm banana plug.