Air density balls
Leverandør: Cornelsen Experimenta
765-0150EA 0 NOK
765-0150
Air density balls
Lærings materiell Læremateriale for fysikk Properties of Matter Learning Activities in Physics
Consists of a ball with check valve, vent valve and suspension hook. For use in conjunction with an electronic scale (0,001 g).
  • Ball diameter: 90 mm