Vacuum pump
Leverandør: EDUCATIONSCIENCE COLLECTION P
765-0317EA 0 NOK
765-0317
Vacuum pump
Pumper Rotasjonsspumper
Two-stage vacuum pump with alternating current motor. Equipped with gas ballast valve to prevent condensation inside the pump.
  • Starting voltage: 0.5 V.