Centrifugal power device with experimental trolley
Leverandør: PHYWE
11008.00 11060.00 03949.00 02206.01 02206.02
765-1382EA 0 NOK
765-1382 765-1383 765-1384 765-1386 765-1385
Centrifugal power device with experimental trolley
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
With the equipment, centrifugal forces that arise during a body's rotation around a specific axis can be measured and examined. The device consists of a plane with a path for a cart. On the underside there is a vertical shaft that is mounted in a bearing. The plane is made to rotate with a laboratory motor connected with a drive belt to the bearing. The experimental trolley is placed on the track and prevented from leaving the track by two bent steel wires. A wire is connected to the trolley that runs over a pulley and on to a dynamometer.