Electric motor, standard
Leverandør: EDUCATIONSCIENCE COLLECTION P
766-0293EA 0 NOK
766-0293
Electric motor, standard
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Very affordable DC motor 1,5 to 4,5 V. Power consumption 0,1 to 0,4 A at speeds 5000 to 6500 min⁻¹. Shaft Ø 2 mm with protrusion 7 mm. Size 24×27 mm. Solder connection.
  • Pack of 10 engines