Planck's constant, experimental equipment
Leverandør: PHYWE
13002.00 11615.05 13626.93 06778.00 08461.00 08463.00 07542.10 08120.14
765-1313EA 2075 NOK
765-1313 765-1310 765-1323 765-1205 765-1226 765-1227 765-1208 765-1211
Planck's constant, experimental equipment
Lærings materiell Læremateriale for fysikk Atomic Physics Learning Activities
Equipment for demonstrating photoelectric power and determining Planck's constant. A photocell is equipped with filters that only transmit light of a certain wavelength. The photocell is illuminated with light from a Hg lamp. The potassium cathode then emits electrons that pass over to the unlit anode. Over time, a voltage arises that prevents new electrons from reaching the anode. The resulting voltage is measured with a measuring amplifier and a voltmeter. The measurement is repeated with filters for different wavelengths. Photocell and Hg lamp are equipped with magnets at the bottom forMore Product Information