Smoke chambers
Leverandør: EDUCATIONSCIENCE COLLECTION P
766-0193EA 0 NOK
766-0193 765-0257
Smoke chambers
Lærings materiell Læremateriale for fysikk Modern Physics Learning Activities
Continuous mist chamber for visualisation of particle traces from alpha and beta preparations. The mist chamber is added a few drops of isopropanol and connected to circulating water flow (<lt />16 °C, flow 1 L/min) and direct voltage (8 to 12 V approx. 8 A).
The process starts after a few minutes and lasts until the isopropanol is consumed.
The fog chamber is equipped with a contrast-rich LED lighting for the best possible results.
Excess interfering ions formed during the process are easily neutralized by wiping the recipient with an antistatic cloth.
Leveringsinformasjon: Tripod rod 12 mm, LED lighting, 100 ml isopropanol and syringe.