Blood pressure / heart rate monitor, fully automatic
Leverandør: EDUCATIONSCIENCE COLLECTION P
763-0225EA 0 NOK
763-0225
Blood pressure / heart rate monitor, fully automatic
Blodtrykks monitor
Wrist meter RX3-Plus.
With integrated cuff and gauge. RX3-Plus measures the pressure during the inflation phase. The meter automatically stores the last 21 measurements in time and date. Also shows an average of the 3 most recent measurement results.