Tripod rods, square, PASS
Leverandør: PHYWE
02026.55 02027.55 02028.55 02029.55
765-1141EA 0 NOK
765-1141 765-1142 765-1143 765-1144
Tripod rods, square, PASS
Stativstang
Tripod rods with square cross section 12×12 mm. Made of galvanized galvanized steel pipe. One end of the rod is provided with a cylindrical rod (50×10 mm). The other end closed. Square rods provide unmatched torsional rigidity compared to cylindrical frame rods.