Defender™ 6000 Coloumn Mount Bench Scales
Leverandør: OHAUS
30575556 30575557 30575558 30575559 30575560 30612499 30612500 30612501 30612502 30612503 30612504 30745943 30745944 30729261 30745777 30745838 30745839
OHAU30575556EA 34250 NOK
OHAU30575556 OHAU30575557 OHAU30575558 OHAU30575559 OHAU30575560 OHAU30612499 OHAU30612500 OHAU30612501 OHAU30612502 OHAU30612503 OHAU30612504 OHAU30745943 OHAU30745944 OHAU30729261 OHAU30745777 OHAU30745838 OHAU30745839
Defender™ 6000 Coloumn Mount Bench Scales
Vekter
The Defender 6000 series extreme washdown bench scale is specifically designed for food, chemical and pharmaceutical applications, including production and packaging.
  • 6-digit, 7-segment white LED, 20 mm high digits
  • 100 to 240 VAC/50/60 Hz universal power supply, hardwired
  • i-DT61XWE indicator
Defender 6000 handles the most demanding applications with 316 stainless steel platform and frame, and hermetically-sealed stainless steel IP69K load cell.
150% overload capacity protection, bar matrix check light, customizable print templates, menu and keyMore Product Information
Bestill nå
SPESIFIKASJONER
Spekter driftstemperatur −10...+40 °C −10...+40 °C −10...+40 °C
Environmentally Preferable
Støpseltype EU plug type UK plug type CH plug type