Cubis® II MCA 225P Ultra-High Resolution Semi-Micro Balances
Leverandør: Sartorius Balances
MCA225P3CE12M MCA225P3S024M MCA225P3S023H MCA225P3S0M MCA225P3S02 MCA225P3CE234I MCA225P3CE99I MCA225P3CE4M MCA225P3CE1I MCA225P3CE34H MCA225P3S014I MCA225P3CE14I MCA225P3S023 MCA225P3CE23 MCA225P3S023I MCA225P3S03H MCA225P3CE2M MCA225P3CE13H MCA225P3CE2 MCA225P3CE4 MCA225P3CE23M MCA225P3S0123H MCA225P3CE14 MCA225P3S034I MCA225P3CE123I MCA225P3CE12H MCA225P3S099M MCA225P3S03I MCA225P3S0123M MCA225P3S034 MCA225P3S0234I MCA225P3CE13 MCA225P3S02H MCA225P3S0234H MCA225P3CE12I MCA225P3S012I MCA225P3CE14M MCA225P3CE3M MCA225P3CE123H MCA225P3CE234H MCA225P3S0124I MCA225P3CE124 MCA225P3CE4H MCA225P3S012H MCA225P3S024I MCA225P3CE4I MCA225P3S04 MCA225P3CE124M MCA225P3S02M MCA225P3CE234M MCA225P3CE24I MCA225P3CE124H MCA225P3CE3I MCA225P3S01I MCA225P3CE14H MCA225P3CE124I MCA225P3CE1M MCA225P3S014H MCA225P3CE123 MCA225P3CE34 MCA225P3CE3 MCA225P3S04H MCA225P3CE2H MCA225P3S034H MCA225P3S013 MCA225P3S01M MCA225P3CE2I MCA225P3CE24H MCA225P3CE13M MCA225P3S03M MCA225P3CE12 MCA225P3S024H MCA225P3S014 MCA225P3S0234 MCA225P3S012M MCA225P3CE3H MCA225P3S04M MCA225P3S04I MCA225P3S03 MCA225P3S034M MCA225P3CE23H MCA225P3S013I MCA225P3CE234 MCA225P3CE24 MCA225P3S099I MCA225P3CEM MCA225P3S024 MCA225P3S013H MCA225P3S0124M MCA225P3S0123I MCA225P3S0124H MCA225P3CE24M MCA225P3CE99M MCA225P3CE123M MCA225P3CE34I MCA225P3S013M MCA225P3S023M MCA225P3S0I MCA225P3S0123 MCA225P3S0124 MCA225P3CEI MCA225P3CE23I MCA225P3CE34M MCA225P3S014M MCA225P3S0234M MCA225P3S02I MCA225P3S012 MCA225P3CE13I
SARBMCA225P3CE12MEA 0 NOK
SARBMCA225P3CE12M SARBMCA225P3S024M SARBMCA225P3S023H SARBMCA225P3S0M SARBMCA225P3S02 SARBMCA225P3CE234I SARBMCA225P3CE99I SARBMCA225P3CE4M SARBMCA225P3CE1I SARBMCA225P3CE34H SARBMCA225P3S014I SARBMCA225P3CE14I SARBMCA225P3S023 SARBMCA225P3CE23 SARBMCA225P3S023I SARBMCA225P3S03H SARBMCA225P3CE2M SARBMCA225P3CE13H SARBMCA225P3CE2 SARBMCA225P3CE4 SARBMCA225P3CE23M SARBMCA225P3S0123H SARBMCA225P3CE14 SARBMCA225P3S034I SARBMCA225P3CE123I SARBMCA225P3CE12H SARBMCA225P3S099M SARBMCA225P3S03I SARBMCA225P3S0123M SARBMCA225P3S034 SARBMCA225P3S0234I SARBMCA225P3CE13 SARBMCA225P3S02H SARBMCA225P3S0234H SARBMCA225P3CE12I SARBMCA225P3S012I SARBMCA225P3CE14M SARBMCA225P3CE3M SARBMCA225P3CE123H SARBMCA225P3CE234H SARBMCA225P3S0124I SARBMCA225P3CE124 SARBMCA225P3CE4H SARBMCA225P3S012H SARBMCA225P3S024I SARBMCA225P3CE4I SARBMCA225P3S04 SARBMCA225P3CE124M SARBMCA225P3S02M SARBMCA225P3CE234M SARBMCA225P3CE24I SARBMCA225P3CE124H SARBMCA225P3CE3I SARBMCA225P3S01I SARBMCA225P3CE14H SARBMCA225P3CE124I SARBMCA225P3CE1M SARBMCA225P3S014H SARBMCA225P3CE123 SARBMCA225P3CE34 SARBMCA225P3CE3 SARBMCA225P3S04H SARBMCA225P3CE2H SARBMCA225P3S034H SARBMCA225P3S013 SARBMCA225P3S01M SARBMCA225P3CE2I SARBMCA225P3CE24H SARBMCA225P3CE13M SARBMCA225P3S03M SARBMCA225P3CE12 SARBMCA225P3S024H SARBMCA225P3S014 SARBMCA225P3S0234 SARBMCA225P3S012M SARBMCA225P3CE3H SARBMCA225P3S04M SARBMCA225P3S04I SARBMCA225P3S03 SARBMCA225P3S034M SARBMCA225P3CE23H SARBMCA225P3S013I SARBMCA225P3CE234 SARBMCA225P3CE24 SARBMCA225P3S099I SARBMCA225P3CEM SARBMCA225P3S024 SARBMCA225P3S013H SARBMCA225P3S0124M SARBMCA225P3S0123I SARBMCA225P3S0124H SARBMCA225P3CE24M SARBMCA225P3CE99M SARBMCA225P3CE123M SARBMCA225P3CE34I SARBMCA225P3S013M SARBMCA225P3S023M SARBMCA225P3S0I SARBMCA225P3S0123 SARBMCA225P3S0124 SARBMCA225P3CEI SARBMCA225P3CE23I SARBMCA225P3CE34M SARBMCA225P3S014M SARBMCA225P3S0234M SARBMCA225P3S02I SARBMCA225P3S012 SARBMCA225P3CE13I
Cubis® II MCA 225P Ultra-High Resolution Semi-Micro Balances
Vekter
The Cubis® II laboratory balances are modular, therefore they allow to choose between applications and configurations which suit the best to the needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions.
  • Draft shield (D) for high capacity micro balances with upgradable hardware
  • Superior weighing performance
  • No more issues with charged samples
  • Cleaning process guidance
  • Hardware upgradeability, compliance and data integrity
  • Easy to use with motorised inner draft shield
MCA display - 7"More Product Information