Natriumtetrahydroborat 98.5-101.5% (gas-volumetric), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Leverandør: Merck
Synonymer: Sodium tetrahydridoborate, Sodium tetrahydroborate
80546-100G 80546-25G
80546-100GEA 2980 NOK
80546-100G 80546-25G
Natriumtetrahydroborat 98.5-101.5% (gas-volumetric), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Sodium borohydride
Formel: NaBH₄
MW: 37,83 g/mol
MDL nummer: MFCD00003518
CAS-nummer: 16940-66-2