SurePac™ Bio 550 SEC MDi™ Size Exclusion Chromatography Columns
Leverandør: Thermo Fisher Scientific
43903-032131 43903-034631 43903-037831 43903-152131 43903-154631 43903-157831
HYPE43903-032131EA 8325 NOK
HYPE43903-032131 HYPE43903-034631 HYPE43903-037831 HYPE43903-152131 HYPE43903-154631 HYPE43903-157831
SurePac™ Bio 550 SEC MDi™ Size Exclusion Chromatography Columns
Kromatografikolonner
SurePac™ Bio 550 SEC MDi™ columns and guards are based on high-purity, spherical, porous (550 Å) monodispersed silica particles that are covalently modified with a proprietary diol hydrophilic layer.
  • Ideal for AAV gene therapy characterisation and quality control assessment due to selectivity of the dedicated smaller particle (3 µm) combined with wide-pore (550 Å) size
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing
  • High-efficiency peaks with excellent recovery
  • Higher
More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Maks. trykk 241 bar
pH område 2 - 8
Temperaturgrense 20...60 °C