Hygienox Electropolished Trolleys
Leverandør: TEKNOMEK
SHTTHYEP2S6949 SHTTHYEP2S8959 SHTTHYEP2S1069 SHTTHYEP3S6949 SHTTHYEP3S8959 SHTTHYEP3S1069
TEKMSHTTHYEP2S6949EA 0 NOK
TEKMSHTTHYEP2S6949 TEKMSHTTHYEP2S8959 TEKMSHTTHYEP2S1069 TEKMSHTTHYEP3S6949 TEKMSHTTHYEP3S8959 TEKMSHTTHYEP3S1069
Hygienox Electropolished Trolleys
Traller
Hygienox electropolished 304-grade stainless steel trolleys are made specifically for cleanroom environments, giving extreme protection against pitting and crevice corrosion caused by harsh clean down routines.
  • Chemically and physically resistant
  • Electropolished 304-grade stainless steel
  • Four quiet castors with Two braked as standard
  • Wall bump guards
Providing protection from corrosive chemicals, such as chlorine and hydrogen peroxide, the Hygienox trolleys are also easier to clean due to the smoother surface of the electropolished stainless steel -More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Materiale Stainless steel 304L Stainless steel 304L Stainless steel 304L
Dimensjon hylle 600×400 mm 800×500 mm 1000×600 mm