FRONTIER™ 5000 Rotors
Leverandør: OHAUS
30314830 30314832 30314836 30314839 30472302 83041238 83041011
OHAU30314830EA 9375 NOK
OHAU30314830 OHAU30314832 OHAU30314836 OHAU30314839 OHAU30472302 OHAU83041238 OHAU83041011
FRONTIER™ 5000 Rotors
Sentrifuger Rotorer
Frontier rotors extend the functionality of Frontier centrifuges by increasing the number of tubes of varied types and sizes that can be used in conjunction with each of the centrifuge models. Easy to clean and operate, the rotors are durably constructed to withstand demanding centrifugation.
  • Aluminium or polypropylene material
  • Easy-installation
  • The sturdy rotors are constructed of high-quality materials and built to last through demanding workflows from autoclave sterilisation to sub-zero temperature
  • Reliable, versatile and ideal for numerous laboratory
More Product Information
Bestill nå
SPESIFIKASJONER
Modell R-A6x50/6MI R-A30x15/4MI R-A30X2/20MIS R-A12X2/30MI R-A24X2/13MB R-A24X2/16MIB R-S-4X200/5MIS
Environmentally Preferable
Maks. kapasitet 6×50 ml 30×15 ml 30×1,5/2 ml 12×1,5/2 ml 24×1,5/2 ml 24×1,5/2 ml 4×200 ml
Maks. Ø for rør (mm) 29 mm/50 ml 18 mm/15 ml 12 mm/2,0 ml, 1,5 ml 12 mm/2,0 ml, 1,5 ml 12 mm/2,0 ml, 1,5 ml 12 mm/2,0 ml, 1,5 ml
Rotor type Angle Angle Angle Angle Angle Angle Swing-out
Vinkel 40° 35° 45° 40° 45° 45° 0 - 90°
Maks. RCF 4427×g 2830×g 42032×g 65395×g 17317×g 24325×g 4136×g
Maks. hastighet 6000 min⁻¹ 4500 min⁻¹ 20000 min⁻¹ 30000 min⁻¹ 13500 min⁻¹ 16000 min⁻¹ 5000 min⁻¹