Hei-PLATE Mix 'n' Heat Core Magnetic Stirrers
Leverandør: Heidolph Instruments GmbH & Co.KG
506-11100-00 506-12100-00
HEID506-11100-00EA 11450 NOK
HEID506-11100-00 HEID506-12100-00
Hei-PLATE Mix 'n' Heat Core Magnetic Stirrers
Omrørere Magnetrørere med varmeplate
In accordance with the motto 'simply stir and heat', our entry-level model Hei-PLATE Mix 'n' Heat Core was designed with two rotary knobs to allow direct access to the two main functions. The speed and temperature are set using a scale. The maximum values are 1400 rpm and 300 °C.
  • Patented Kera-Disk® footprint made of anodised aluminum
  • Chemical-resistant
  • Scratch-proof
  • Has a diameter of 145 mm
This magnetic stirrer is very easy to use and is equipped with the most important safety features. The large safety distance between the heating plate and theMore Product Information
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Platemateriale Kera-Disk®, keramisk belagt silumin (aluminiumslegering) Kera-Disk®, keramisk belagt silumin (aluminiumslegering)
Platemål 145 mm 135 mm
Total dimensjon 168×299×101 mm 168×299×101 mm
Elektronisk 230 V, 50/60 Hz 230 V, 50/60 Hz
Vekt 3 kg 3 kg