VitraPOR® Filter Discs, Biplane, Series 16, Porosity Fine
Leverandør: ROBU
VitraPOR®
ROBU6140FEA 6750 NOK
ROBU6140F ROBU6130F ROBU1690F ROBU1680F ROBU1670F ROBU1620F ROBU1610F ROBU1611F ROBU1612F ROBU1660F ROBU1650F ROBU1640F ROBU1630F ROBU1625F ROBU6200F ROBU6190F ROBU6180F ROBU6170F ROBU6160F ROBU6150F
VitraPOR® Filter Discs, Biplane, Series 16, Porosity Fine
Filter Filterdisker
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for use in many applications.
  • Very good chemical resistance
  • Very good temperature resistance
  • Porosity: 4 to 5,5 µm
Made of borosilicate 3.3 glass. The faces of filter discs from the series 16 are precisely ground to tight tolerances and plane surfaces as may be required in specific installation circumstances.