2-Aminoisobutyric acid 99%
Leverandør: Thermo Scientific
Synonymer: α-Aminoisobutyric acid, 2-Amino-2-methylpropionic acid, 2-Metilalanina, H-Aib-OH, 2-Methylalanine, 2-Amino-2-methylpropanoic acid
A13021.14 A13021.22 A13021.30
A13021.14EA 630 NOK
A13021.14 A13021.22 A13021.30
2-Aminoisobutyric acid 99%
2-Aminoisobutyric acid
Purity: 99% CAS No.: 62-57-7 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Melting Point: 300° Beilstein Reference: 506496 Merck Reference: 14,445 MDL No.: MFCD00008049
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Kokepunkt: 204 °C (1013 hPa)
Smeltepunkt: >300 °C
Tetthet: 1,278 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008049
CAS-nummer: 62-57-7
EINECS: 200-544-0
Merck Indeks: 14,445