SC-514 ≥98% (by TLC)
Leverandør: ENZO LIFE SCIENCES
Synonymer: 4-amino-[2,3'-bithiophene]-5-carboxamide, IKK-2 Inhibitor, SC-514
BMLEI3430010 BMLEI3430050
ENZOBMLEI3430010EA 1300 NOK
ENZOBMLEI3430010 ENZOBMLEI3430050
SC-514 ≥98% (by TLC)
SC-514
IKK inhibitor
Selective, reversible and ATP-competitive inhibitor of IKK2 (IC50=11.2µM). Does not inhibit other IKK isoforms or other serine/threonine or tyrosine kinases. Inhibits transcription of NF-κB dependent genes in IL-1β-induced rheumatoid arthritis-derived synovial fibroblasts in a dose-dependent fashion.
Formel: C₉H₈N₂OS₂
MW: 224.3 g/mol
MDL nummer: MFCD00206459
CAS-nummer: 354812-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce30194d2fd74e36af9da9677d3d85d828d2f2fb&vid=efb1fa57280b1a664b5e5f677e839aa7b4ac4358&prid=web&clid=SAPDAM