4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥98% (by HPLC)
Leverandør: ENZO LIFE SCIENCES
Synonymer: 4-(Dimethylamino)-N-(7-(hydroxyamino)-7-oxoheptyl)benzamide, M344, M 344
ALX270297M001 ALX270297M005
ENZOALX270297M001EA 2025 NOK
ENZOALX270297M001 ENZOALX270297M005
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide ≥98% (by HPLC)
4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
Potent inhibitor of histone deacetylases (HDACs) (IC50≤1µM).
  • HDAC inhibitor
White crystalline solid.
Soluble in DMSO (1mg/ml)
Formel: C₁₆H₂₅N₃O₃
MW: 307,39 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD03453554
CAS-nummer: 251456-60-7