Nutlin-3 ≥98% (by HPLC)
Leverandør: ENZO LIFE SCIENCES
Synonymer: 4-(cis-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one, (±)-4-[4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4,5-dihydro-imidazole-1-carbonyl]-piperazin-2-once
ALX430128M001 ALX430128M005 ALX430128M025
ENZOALX430128M001EA 815 NOK
ENZOALX430128M001 ENZOALX430128M005 ENZOALX430128M025
Nutlin-3 ≥98% (by HPLC)
Nutlin-3
Inhibitor of p53/MDM2 interaction
(+/-) -Nutlin-3 is an inhibitor of p53/MDM2 interaction (IC50=0.09µM).
Formel: C₃₀H₃₀Cl₂N₄O₄
MW: 581,49 g/mol
MDL nummer: MFCD07784509
CAS-nummer: 548472-68-0