Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 532)

Leverandør: AAT BIOQUEST
10280070 10280071
AATB10280070EA 4350 NOK
AATB10280070 AATB10280071
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 532)
Antistoffer
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR