Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 810)

Leverandør: AAT BIOQUEST
116100O0 116100O1
AATB116100O0EA 4350 NOK
AATB116100O0 AATB116100O1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 810)
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 810
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR