Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Violet 500)

Leverandør: AAT BIOQUEST
11610100 11610101
AATB11610100EA 4350 NOK
AATB11610100 AATB11610101
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (mFluor™ Violet 500)
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Affinity purified
  • Easily reconstituted

NB: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR