Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE/iFluor® 594)

Leverandør: AAT BIOQUEST
100811Y0 100811Y1 100811Y2
AATB100811Y0EA 3750 NOK
AATB100811Y0 AATB100811Y1 AATB100811Y2
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (PE/iFluor® 594)
Antistoffer
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR