Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (TRITC)

Leverandør: AAT BIOQUEST
100811I0 100811I1
AATB100811I0EA 4350 NOK
AATB100811I0 AATB100811I1
Anti-CD8a Mouse Monoclonal Antibody [clone: SK1] (TRITC)
Antistoffer
SK1 is an anti-human monoclonal antibody that is specific for the CD8a antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: SK1
Conjugation: TRITC
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD8a CD8a
Environmentally Preferable
Antigen-navn CD8a antigen CD8a antigen
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate) TRITC (Tetramethylrhodamine Isothiocyanate)
Klon SK1 SK1
Reaktivitet Human
Vert Mouse Mouse
Isotype
ImmunoFluorescens Yes Yes
Flowcytometri Yes Yes
Rensing Affinity chromatography Affinity chromatography
Kyssabsorpsjon No No
Lagringstemp 2...8 °C with minimised light exposure, do not freeze 2...8 °C with minimised light exposure, do not freeze
Frakttemp. 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR