Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 750)

Leverandør: AAT BIOQUEST
101920L0 101920L1
AATB101920L0EA 4350 NOK
AATB101920L0 AATB101920L1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (iFluor® 750)
Antistoffer
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: iFluor® 750
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR