Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 633)

Leverandør: AAT BIOQUEST
103800E0 103800E1
AATB103800E0EA 4350 NOK
AATB103800E0 AATB103800E1
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 633)
Antistoffer
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 633
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol CD38 CD38
Environmentally Preferable
Antigen-navn CD38 antigen CD38 antigen
Antistofftype Primary Primary
Klonalitet Monoclonal Monoclonal
Konjugering iFluor® 633 iFluor® 633
Klon HIT2 HIT2
Reaktivitet Human
Vert Mouse Mouse
Isotype IgG1 IgG1
ImmunoFluorescens Yes Yes
Flowcytometri Yes Yes
Rensing Affinity chromatography Affinity chromatography
Kyssabsorpsjon No No
Lagringstemp Refrigerated (2...8 °C), minimise light exposure Refrigerated (2...8 °C), minimise light exposure
Frakttemp. 20 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR