Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (mFluor™ Violet 500)

Leverandør: AAT BIOQUEST
10191100 10191101
AATB10191100EA 4350 NOK
AATB10191100 AATB10191101
Anti-CD19 Mouse Monoclonal Antibody [clone: SJ25C1] (mFluor™ Violet 500)
Antistoffer
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.

  • Affinity purified
  • Easily reconstituted

NB: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR