Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE (Phycoerythrin)/iFluor® 700)

Leverandør: AAT BIOQUEST INC
116101X0 116101X1 116101X2
AATB116101X0EA 3750 NOK
AATB116101X0 AATB116101X1 AATB116101X2
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PE (Phycoerythrin)/iFluor® 700)
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: PE (Phycoerythrin)/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR