Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: AAT BIOQUEST
116101I0 116101I1
AATB116101I0EA 4350 NOK
AATB116101I0 AATB116101I1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

NB: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR