Anti-CD25 Rat Monoclonal Antibody [clone: 3C7] (PE)

Leverandør: STEMCELL Technologies
60010PE 60010PE.1
STMC60010PEEA 0 NOK
STMC60010PE STMC60010PE.1
Anti-CD25 Rat Monoclonal Antibody [clone: 3C7] (PE)
Antistoffer
Rat monoclonal IgG2b antibody against mouse CD25, PE-conjugated.

  • T cells
  • T cells Regulatory

The 3C7 antibody reacts with murine CD25 (interleukin‐2 receptor α chain; IL‐2Rα), an ~55 kDa type 1 transmembrane glycoprotein expressed on the surface of T and B cell progenitors and activated mature B and T cells. CD25 has been used as a marker to identify CD4+/FoxP3+ regulatory T cells in mice. Mutations in CD25 result in severe immunodeficiency, underscoring the importance of the roles the protein plays in the differentiation, activation and proliferationMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn CD25
Environmentally Preferable
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)
Klon 3C7
Reaktivitet Mouse
Vert Rat
Gen-ID 16184
Isotype IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR