Anti-IgG Goat Polyclonal Antibody (XFD750)

Leverandør: AAT BIOQUEST
50180 50181
AATB50180EA 1425 NOK
AATB50180 AATB50181
Anti-IgG Goat Polyclonal Antibody (XFD750)
Antistoffer
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.

  • Affinity purified
  • Easily reconstituted

Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).

NB: For research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: XFD750
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG
Environmentally Preferable
Antigen-navn Immunoglobulin G
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering XFD750
Reaktivitet Human
Vert Goat
Isotype IgG (H, L)
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Kyssabsorpsjon No
Lagringstemp Freeze (<–15 °C); Minimise light exposure
Frakttemp. 20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR