Anti-KLH Mouse Monoclonal Antibody [clone: KT119]

Leverandør: ANTIBODIES.COM
A133-100 A133-200
ANTIA133-100EA 5475 NOK
ANTIA133-100 ANTIA133-200
Anti-KLH Mouse Monoclonal Antibody [clone: KT119]
Antistoffer
Mouse monoclonal (KT119) antibody to KLH for isotype control.

  • Validated applications: Isotype control
  • Host: Mouse
  • Clonality: Monoclonal
  • Isotype: IgG2b
  • Conjugate: Unconjugated

Type: Primary
Antigen: KLH
Clonality: Monoclonal
Clone: KT119
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR