ReadiUse™ 100 bp DNA Ladder
Leverandør: AAT BIOQUEST
60070
60071
AATB60070EA
1550
NOK
AATB60070
AATB60071
ReadiUse™ 100 bp DNA Ladder
Elektroforese reagenser
Molekylvekt markører
ReadiUse™ 100 bp DNA ladder is a ready-to-use DNA ladder, which is pre-mixed with loading dye for direct gel loading.
- Stable for 12 months
Quality tested in-house.
NB: For research use only.